COMH4211A946 Combi-Blocks, Inc.

3-Bromo-4-methoxy-n-((1-methyl-1h-pyrazol-4-yl)methyl)benzamide

  • MDL number MFCD13430804
  • Cas Number : 1305756-18-6
  • Molecular Weight : 324.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-4-methoxy-N-((1-methyl-1H-pyrazol-4-yl)methyl)benzamide
Inchi Code 1S/C13H14BrN3O2/c1-17-8-9(7-16-17)6-15-13(18)10-3-4-12(19-2)11(14)5-10/h3-5,7-8H,6H2,1-2H3,(H,15,18)
Inchi Key UJNJJLIIJWOCCJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0252

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0252
Availability details for
Quantity
  • Availability
  • Add to Cart