COMH4211A948 Combi-Blocks, Inc.

3-(5-Fluoro-2-trifluoromethyl-phenyl)-2-methyl-propionic acid

  • MDL number MFCD31579328
  • Cas Number : 2229270-83-9
  • Molecular Weight : 250.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0257

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0257
Availability details for
Quantity
  • Availability
  • Add to Cart