COMH4211A94C Combi-Blocks, Inc.

6-(2,2,2-Trifluoro-ethoxy)-1h-indazol-3-ylamine

  • MDL number MFCD31381590
  • Cas Number : 2169618-52-2
  • Molecular Weight : 231.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0261
Availability details for
Quantity
  • Availability
  • Add to Cart