COMH4211A950 Combi-Blocks, Inc.

4-Amino-3-(benzyloxy)-n-methylbenzamide

  • MDL number MFCD14559373
  • Cas Number : 1216112-24-1
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0272

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0272
Availability details for
Quantity
  • Availability
  • Add to Cart