COMH4211A951 Combi-Blocks, Inc.

N1,N2-Dipropylethanediamide

  • MDL number MFCD00027894
  • Cas Number : 14040-77-8
  • Molecular Weight : 172.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0274

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0274
Availability details for
Quantity
  • Availability
  • Add to Cart