COMH4211A952 Combi-Blocks, Inc.

Methyl 4-(1-hydroxyethyl)-2-methylbenzoate

  • MDL number MFCD30094554
  • Cas Number : 1823311-32-5
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0276

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0276
Availability details for
Quantity
  • Availability
  • Add to Cart