COMH4211A953 Combi-Blocks, Inc.

1-(2-Aminoethyl)-3-benzylurea hydrochloride

  • MDL number MFCD30486309
  • Cas Number : 646536-50-7
  • Molecular Weight : 229.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0279

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0279
Availability details for
Quantity
  • Availability
  • Add to Cart