COMH4211A954 Combi-Blocks, Inc.

3-Bromo-4-(cyclopropylmethoxy)benzaldehyde

  • MDL number MFCD17284328
  • Cas Number : 628711-48-8
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0281

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0281
Availability details for
Quantity
  • Availability
  • Add to Cart