COMH4211A955 Combi-Blocks, Inc.

2(1H)-Pyridinone, 4-[(2,4-difluorophenyl)methoxy]-6-methyl-

  • MDL number MFCD31613925
  • Cas Number : 586379-20-6
  • Molecular Weight : 251.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0282

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0282
Availability details for
Quantity
  • Availability
  • Add to Cart