COMH4211A95D Combi-Blocks, Inc.

2-[3-Carboxy-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic acid

  • MDL number MFCD29050077
  • Cas Number : 1239233-87-4
  • Molecular Weight : 335.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0300

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0300
Availability details for
Quantity
  • Availability
  • Add to Cart