COMH4211A95E Combi-Blocks, Inc.

1-Dimethylamino-cyclopropanecarboxylic acid hydrochloride

  • MDL number MFCD31557912
  • Cas Number : 1049717-21-6
  • Molecular Weight : 165.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0301

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0301
Availability details for
Quantity
  • Availability
  • Add to Cart