COMH4211A95F Combi-Blocks, Inc.

N-(3-Bromopropyl)-4-trifluoromethylnicotinamide

  • MDL number MFCD31539520
  • Cas Number : 750592-58-6
  • Molecular Weight : 311.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0302

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0302
Availability details for
Quantity
  • Availability
  • Add to Cart