COMH4211A960 Combi-Blocks, Inc.

3-Bromo-4-(2,4-difluoro-phenoxy)-benzylamine

  • MDL number MFCD25801003
  • Cas Number : 1549154-52-0
  • Molecular Weight : 314.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0307

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0307
Availability details for
Quantity
  • Availability
  • Add to Cart