COMH4211A961 Combi-Blocks, Inc.

(2R)-1-(Ethanesulfonyl)-2-methylpiperazine

  • MDL number MFCD24301062
  • Cas Number : 1568167-85-0
  • Molecular Weight : 192.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-1-(ethylsulfonyl)-2-methylpiperazine
Inchi Code 1S/C7H16N2O2S/c1-3-12(10,11)9-5-4-8-6-7(9)2/h7-8H,3-6H2,1-2H3/t7-/m1/s1
Inchi Key BOSBMDOCEQZSDP-SSDOTTSWSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0308

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0308
Availability details for
Quantity
  • Availability
  • Add to Cart