COMH4211A962 Combi-Blocks, Inc.

S-(2-Hydroxy-1-methyl-ethyl)-carbamic acid allyl ester

  • MDL number MFCD21308333
  • Cas Number : 1567958-79-5
  • Molecular Weight : 159.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0309

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0309
Availability details for
Quantity
  • Availability
  • Add to Cart