COMH4211A963 Combi-Blocks, Inc.

1-Cyclopropyl-n-(thiophen-2-ylmethyl)ethanamine

  • MDL number MFCD07352692
  • Cas Number : 871805-58-2
  • Molecular Weight : 181.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0311

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0311
Availability details for
Quantity
  • Availability
  • Add to Cart