COMH4211A964 Combi-Blocks, Inc.

4-Bromo-3-(4,4-difluoro-piperidin-1-ylmethyl)-phenylamine

  • MDL number MFCD31614013
  • Cas Number : 2270907-74-7
  • Molecular Weight : 305.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0314

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0314
Availability details for
Quantity
  • Availability
  • Add to Cart