COMH4211A966 Combi-Blocks, Inc.

(1-Ethyl-1h-pyrazol-4-yloxy)-acetic acid

  • MDL number MFCD27439128
  • Cas Number : 1592704-22-7
  • Molecular Weight : 170.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0322

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0322
Availability details for
Quantity
  • Availability
  • Add to Cart