COMH4211A968 Combi-Blocks, Inc.

3-Amino-4-fluoro-n-methoxybenzamide

  • MDL number MFCD11110231
  • Cas Number : 348165-47-9
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0324

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0324
Availability details for
Quantity
  • Availability
  • Add to Cart