COMH4211A96A Combi-Blocks, Inc.

4-Chloro-6-(2,2,2-trifluoroethoxy)pyrimidine

  • MDL number MFCD14611084
  • Cas Number : 155957-47-4
  • Molecular Weight : 212.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0327

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0327
Availability details for
Quantity
  • Availability
  • Add to Cart