COMH4211A96D Combi-Blocks, Inc.

4-Amino-bicyclo[2.2.2]octane-1-carboxylic acid methylamide, hydrochloride

  • MDL number MFCD31539553
  • Cas Number : 2206607-66-9
  • Molecular Weight : 218.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0335

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0335
Availability details for
Quantity
  • Availability
  • Add to Cart