COMH4211A96E Combi-Blocks, Inc.

3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(2,2,2-trifluoroethyl)-1h-pyrazole

  • MDL number MFCD30745247
  • Molecular Weight : 304.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0336

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0336
Availability details for
Quantity
  • Availability
  • Add to Cart