COMH4211A96F Combi-Blocks, Inc.

(4-Hydroxy-butyl)-methyl-carbamic acid benzyl ester

  • MDL number MFCD16621755
  • Cas Number : 116644-44-1
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 4-hydroxybutyl(methyl)carbamate
Inchi Code 1S/C13H19NO3/c1-14(9-5-6-10-15)13(16)17-11-12-7-3-2-4-8-12/h2-4,7-8,15H,5-6,9-11H2,1H3
Inchi Key QGKLOOPAWNEWQM-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0337

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0337
Availability details for
Quantity
  • Availability
  • Add to Cart