COMH4211A972 Combi-Blocks, Inc.

5-Bromo-n-cyclopropyl-2-(2-methoxy-ethoxy)-benzamide

  • MDL number MFCD31628802
  • Cas Number : 2270911-10-7
  • Molecular Weight : 314.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0341

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0341
Availability details for
Quantity
  • Availability
  • Add to Cart