COMH4211A976 Combi-Blocks, Inc.

Methanone, (4-bromo-2-methylphenyl)-4-morpholinyl-

  • MDL number MFCD21369817
  • Cas Number : 276677-17-9
  • Molecular Weight : 284.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0350

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0350
Availability details for
Quantity
  • Availability
  • Add to Cart