COMH4211A978 Combi-Blocks, Inc.

1-(3-Bromo-4-propoxy-phenyl)-ethylamine

  • MDL number MFCD17247770
  • Cas Number : 1344343-86-7
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0360

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0360
Availability details for
Quantity
  • Availability
  • Add to Cart