COMH4211A979 Combi-Blocks, Inc.

[1-(Propane-2-sulfonyl)-piperidin-4-yl]-methanol

  • MDL number MFCD12772391
  • Cas Number : 1178653-66-1
  • Molecular Weight : 221.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(isopropylsulfonyl)-4-piperidinyl]methanol
Inchi Code 1S/C9H19NO3S/c1-8(2)14(12,13)10-5-3-9(7-11)4-6-10/h8-9,11H,3-7H2,1-2H3
Inchi Key QIAVJUJMJDFAML-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0361

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0361
Availability details for
Quantity
  • Availability
  • Add to Cart