COMH4211A97A Combi-Blocks, Inc.

(2-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenoxy]-ethyl)-carbamic acid tert-butyl ester

  • MDL number MFCD31579498
  • Cas Number : 1505515-74-1
  • Molecular Weight : 363.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0362
Availability details for
Quantity
  • Availability
  • Add to Cart