COMH4211A97B Combi-Blocks, Inc.

(3Ar,5s,6r,6ar)-5-(iodomethyl)-2,2-dimethyl-tetrahydro-2h-furo[2,3-d][1,3]dioxol-6-ol

  • MDL number MFCD24842633
  • Cas Number : 50600-39-0
  • Molecular Weight : 300.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0366

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0366
Availability details for
Quantity
  • Availability
  • Add to Cart