COMH4211A97D Combi-Blocks, Inc.

(3-Methoxy-2-propoxybenzyl)methylamine

  • MDL number MFCD11607676
  • Cas Number : 709649-53-6
  • Molecular Weight : 209.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0372

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0372
Availability details for
Quantity
  • Availability
  • Add to Cart