COMH4211A982 Combi-Blocks, Inc.

2-(5-Phenylthiazol-2-yl)aniline

  • MDL number MFCD16539441
  • Cas Number : 96303-85-4
  • Molecular Weight : 252.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0385

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0385
Availability details for
Quantity
  • Availability
  • Add to Cart