COMH4211A983 Combi-Blocks, Inc.

5-Bromo-2-chloro-n-(2-methylpropyl)pyridine-3-carboxamide

  • MDL number MFCD16159743
  • Cas Number : 1248384-68-0
  • Molecular Weight : 291.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0386

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0386
Availability details for
Quantity
  • Availability
  • Add to Cart