COMH4211A984 Combi-Blocks, Inc.

Methyl 2-(cyclobutylamino)-4,5-difluorobenzoate

  • MDL number MFCD31613845
  • Cas Number : 1593887-32-1
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0387

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0387
Availability details for
Quantity
  • Availability
  • Add to Cart