COMH4211A986 Combi-Blocks, Inc.

4-Bromo-n-(2-pyrrolidin-1-yl-ethyl)-benzamide

  • MDL number MFCD03785514
  • Cas Number : 796888-28-3
  • Molecular Weight : 297.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0394

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0394
Availability details for
Quantity
  • Availability
  • Add to Cart