COMH4211A989 Combi-Blocks, Inc.

1-Bromo-4-(2-methylsulfanyl-ethoxy)-benzene

  • MDL number MFCD14644617
  • Cas Number : 166959-48-4
  • Molecular Weight : 247.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromophenyl 2-(methylsulfanyl)ethyl ether
Inchi Code 1S/C9H11BrOS/c1-12-7-6-11-9-4-2-8(10)3-5-9/h2-5H,6-7H2,1H3
Inchi Key WOQLPFZINZLWGE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0399

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0399
Availability details for
Quantity
  • Availability
  • Add to Cart