COMH4211A98B Combi-Blocks, Inc.

2-Amino-5-fluoro-n-isopropylbenzamide

  • MDL number MFCD11132587
  • Cas Number : 1094290-34-2
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0404

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0404
Availability details for
Quantity
  • Availability
  • Add to Cart