COMH4211A98C Combi-Blocks, Inc.

2-(2-Nitrobenzylthio)-5-chlorobenzenamine

  • MDL number MFCD12445285
  • Cas Number : 1019455-02-7
  • Molecular Weight : 294.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0407
Availability details for
Quantity
  • Availability
  • Add to Cart