COMH4211A98E Combi-Blocks, Inc.

5-Bromo-2-morpholin-4-yl-nicotinic acid tert-butyl ester

  • MDL number MFCD31557783
  • Cas Number : 2204052-09-3
  • Molecular Weight : 343.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0409

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0409
Availability details for
Quantity
  • Availability
  • Add to Cart