COMH4211A98F Combi-Blocks, Inc.

3-Hydroxy-4-fluoro-n-cyclopropylbenzamide

  • MDL number MFCD23110722
  • Cas Number : 639858-61-0
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0411

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0411
Availability details for
Quantity
  • Availability
  • Add to Cart