COMH4211A992 Combi-Blocks, Inc.

(5-Chloro-pyrazin-2-yl)-piperidin-1-yl-methanone

  • MDL number MFCD31381730
  • Cas Number : 1562188-93-5
  • Molecular Weight : 225.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0420
Availability details for
Quantity
  • Availability
  • Add to Cart