COMH4211A993 Combi-Blocks, Inc.

N-Benzyl-6-hydrazino-nicotinamide

  • MDL number MFCD17950098
  • Cas Number : 1284017-83-9
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0421

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0421
Availability details for
Quantity
  • Availability
  • Add to Cart