COMH4211A995 Combi-Blocks, Inc.

[3-Hydroxymethyl-5-(2,2,2-trifluoro-ethoxy)-phenyl]-methanol

  • MDL number MFCD31628818
  • Cas Number : 2270908-23-9
  • Molecular Weight : 236.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0428

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0428
Availability details for
Quantity
  • Availability
  • Add to Cart