COMH4211A996 Combi-Blocks, Inc.

5-Bromo-2-(cyclopropylmethoxy)benzaldehyde

  • MDL number MFCD12138838
  • Cas Number : 1156170-18-1
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0429

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0429
Availability details for
Quantity
  • Availability
  • Add to Cart