COMH4211A997 Combi-Blocks, Inc.

5-tert-Butoxycarbonylmethoxy-2-hydroxy-benzoic acid methyl ester

  • MDL number MFCD31618899
  • Cas Number : 2270907-86-1
  • Molecular Weight : 282.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0430

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0430
Availability details for
Quantity
  • Availability
  • Add to Cart