COMH4211A998 Combi-Blocks, Inc.

5-Fluoro-4'-methoxybiphenyl-2-carbonitrile

  • MDL number MFCD31579470
  • Cas Number : 1260389-32-9
  • Molecular Weight : 227.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-4'-methoxy-[1,1'-biphenyl]-2-carbonitrile
Inchi Code 1S/C14H10FNO/c1-17-13-6-3-10(4-7-13)14-8-12(15)5-2-11(14)9-16/h2-8H,1H3
Inchi Key VYZOICPJTKGVSM-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0433

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0433
Availability details for
Quantity
  • Availability
  • Add to Cart