COMH4211A999 Combi-Blocks, Inc.

1-(2-Fluoro-benzyl)-3,5-dimethyl-1h-pyrazol-4-ol

  • MDL number MFCD31583226
  • Cas Number : 2270906-03-9
  • Molecular Weight : 220.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0437

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0437
Availability details for
Quantity
  • Availability
  • Add to Cart