COMH4211A99A Combi-Blocks, Inc.

2-Cyclobutylamino-isonicotinic acid methyl ester

  • MDL number MFCD31613871
  • Cas Number : 1697458-71-1
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0439
Availability details for
Quantity
  • Availability
  • Add to Cart