COMH4211A99B Combi-Blocks, Inc.

2-(Oxan-4-ylmethyl)butanoic acid

  • MDL number MFCD16745863
  • Cas Number : 1247454-31-4
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0442
Availability details for
Quantity
  • Availability
  • Add to Cart