COMH4211A99C Combi-Blocks, Inc.

N-(2-(2-(2-Aminoethoxyl)ethoxy)ethyl)-3-(1,3-dioxoisoindolin-2-yl)propanamide

  • MDL number MFCD31579402
  • Cas Number : 1429293-22-0
  • Molecular Weight : 349.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0443

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0443
Availability details for
Quantity
  • Availability
  • Add to Cart