COMH4211A99D Combi-Blocks, Inc.

1-(3-Fluoro-4-methylphenyl)propan-1-amine hydrochloride

  • MDL number MFCD30180578
  • Cas Number : 1955514-22-3
  • Molecular Weight : 203.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0447
Availability details for
Quantity
  • Availability
  • Add to Cart